Merck
  • Solution-processed PbS quantum dot infrared photodetectors and photovoltaics.
Solution-processed PbS quantum dot infrared photodetectors and photovoltaics.
Nature materials (2005-01-11)
Steven A McDonald, Gerasimos Konstantatos, Shiguo Zhang, Paul W Cyr, Ethan J D Klem, Larissa Levina, Edward H Sargent
ABSTRACT
In contrast to traditional semiconductors, conjugated polymers provide ease of processing, low cost, physical flexibility and large area coverage. These active optoelectronic materials produce and harvest light efficiently in the visible spectrum. The same functions are required in the infrared for telecommunications (1,300-1,600 nm), thermal imaging (1,500 nm and beyond), biological imaging (transparent tissue windows at 800 nm and 1,100 nm), thermal photovoltaics (>1,900 nm), and solar cells (800-2,000 nm). Photoconductive polymer devices have yet to demonstrate sensitivity beyond approximately 800 nm (refs 2,3). Sensitizing conjugated polymers with infrared-active nanocrystal quantum dots provides a spectrally tunable means of accessing the infrared while maintaining the advantageous properties of polymers. Here we use such a nanocomposite approach in which PbS nanocrystals tuned by the quantum size effect sensitize the conjugated polymer poly[2-methoxy-5-(2'-ethylhexyloxy-p-phenylenevinylene)] (MEH-PPV) into the infrared. We achieve, in a solution-processed device and with sensitivity far beyond 800 nm, harvesting of infrared-photogenerated carriers and the demonstration of an infrared photovoltaic effect. We also make use of the wavelength tunability afforded by the nanocrystals to show photocurrent spectra tailored to three different regions of the infrared spectrum.
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 1400 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 1200 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 1000 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 1600 nm, 10 mg/mL in toluene