Merck
  • Lipidomics reveals multiple pathway effects of a multi-components preparation on lipid biochemistry in ApoE*3Leiden.CETP mice.
Lipidomics reveals multiple pathway effects of a multi-components preparation on lipid biochemistry in ApoE*3Leiden.CETP mice.
PloS one (2012-02-01)
Heng Wei, Chunxiu Hu, Mei Wang, Anita M van den Hoek, Theo H Reijmers, Suzan Wopereis, Jildau Bouwman, Raymond Ramaker, Henrie A A J Korthout, Marco Vennik, Thomas Hankemeier, Louis M Havekes, Renger F Witkamp, Elwin R Verheij, Guowang Xu, Jan van der Greef
ABSTRACT
Causes and consequences of the complex changes in lipids occurring in the metabolic syndrome are only partly understood. Several interconnected processes are deteriorating, which implies that multi-target approaches might be more successful than strategies based on a limited number of surrogate markers. Preparations from Chinese Medicine (CM) systems have been handed down with documented clinical features similar as metabolic syndrome, which might help developing new intervention for metabolic syndrome. The progress in systems biology and specific animal models created possibilities to assess the effects of such preparations. Here we report the plasma and liver lipidomics results of the intervention effects of a preparation SUB885C in apolipoprotein E3 Leiden cholesteryl ester transfer protein (ApoE*3Leiden.CETP) mice. SUB885C was developed according to the principles of CM for treatment of metabolic syndrome. The cannabinoid receptor type 1 blocker rimonabant was included as a general control for the evaluation of weight and metabolic responses. ApoE*3Leiden.CETP mice with mild hypercholesterolemia were divided into SUB885C-, rimonabant- and non-treated control groups. SUB885C caused no weight loss, but significantly reduced plasma cholesterol (-49%, p<0.001), CETP levels (-31%, p<0.001), CETP activity (-74%, p<0.001) and increased HDL-C (39%, p<0.05). It influenced lipidomics classes of cholesterol esters and triglycerides the most. Rimonabant induced a weight loss (-9%, p<0.05), but only a moderate improvement of lipid profiles. In vitro, SUB885C extract caused adipolysis stimulation and adipogenesis inhibition in 3T3-L1 cells. SUB885C, a multi-components preparation, is able to produce anti-atherogenic changes in lipids of the ApoE*3Leiden.CETP mice, which are comparable to those obtained with compounds belonging to known drugs (e.g. rimonabant, atorvastatin, niacin). This study successfully illustrated the power of lipidomics in unraveling intervention effects and to help finding new targets or ingredients for lifestyle-related metabolic abnormality.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
CETP Activity Assay Kit, Supplied by Roar Biomedical, Inc.
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Avanti
13:0 Lyso LPC, Avanti Research - A Croda Brand
Sigma-Aldrich
Tripentadecanoin, ≥99%