Merck
  • The 1,2-benzenedithiole-based cyclocondensation assay: a valuable tool for the measurement of chemopreventive isothiocyanates.
The 1,2-benzenedithiole-based cyclocondensation assay: a valuable tool for the measurement of chemopreventive isothiocyanates.
Critical reviews in food science and nutrition (2012-03-29)
Yuesheng Zhang
ABSTRACT
Many naturally occurring isothiocyanates (ITCs) show highly promising chemopreventive activities. Humans are commonly exposed to these compounds through the consumption of cruciferous vegetables which are the main source of dietary ITCs. Dietary ITCs may play an important role in cancer prevention and in the well-recognized cancer preventive activities of cruciferous vegetables. A generic analytical method, namely the 1,2-benzenedithiol-based cyclocondensation assay, was previously developed for quantitation of ITCs and their in vivo metabolites. This method has been widely used and has contributed greatly to research on chemoprevention by ITCs. In this article, the discovery and development of the cyclocondensation assay are recapitulated, and its sensitivity and specificity as well as its advantages and limitations are scrutinized. Moreover, detailed discussion is also provided to show how this assay has been used to advance our understanding of the cancer chemopreventive potential and the mechanism of action of ITCs.
Sigma-Aldrich
Benzene-1,2-dithiol, 96%