Merck
  • Isolation and characterization of a new advanced glycation endproduct of dehydroascorbic acid and lysine.
Isolation and characterization of a new advanced glycation endproduct of dehydroascorbic acid and lysine.
Biochimica et biophysica acta (2003-02-22)
Ognyan K Argirov, Bin Lin, Paul Olesen, Beryl J Ortwerth
ABSTRACT
Proteins are subject of posttranslational modification by sugars and their degradation products in vivo. The process is often referred as glycation. L-Dehydroascorbic acid (DHA), an oxidation product of L-ascorbic acid (vitamin C), is known as a potent glycation agent. A new product of modification of lysine epsilon -amino group by DHA was discovered as a result of the interaction between Boc-Lys and dehydroascorbic acid. The chromatographic and spectral analyses revealed that the structure of the product was 1-(5-ammonio-5-carboxypentyl)-3-oxido-4-(hydroxymethyl)pyridinium. The same compound was isolated from DHA modified calf lens protein after hydrolysis and chromatographic separation. The study confirmed that L-erythrulose is an important intermediate of modification of proteins by DHA. The structure of the reported product and in vitro experiments suggested that L-erythrulose could further transform to L-threose, L-erythrose and glycolaldehyde under conditions similar to physiological. The present study revealed that the modification of epsilon -amino groups of lysine residues by DHA is a complex process and could involve a number of reactive carbonyl species.
Sigma-Aldrich
H-Lys(Boc)-OH, ≥95%