Merck
  • Regulation of K(+) current in human airway epithelial cells by exogenous and autocrine adenosine.
Regulation of K(+) current in human airway epithelial cells by exogenous and autocrine adenosine.
American journal of physiology. Cell physiology (2001-11-08)
A J Szkotak, A M Ng, J Sawicka, S A Baldwin, S F Man, C E Cass, J D Young, M Duszyk
ABSTRACT
The regulatory actions of adenosine on ion channel function are mediated by four distinct membrane receptors. The concentration of adenosine in the vicinity of these receptors is controlled, in part, by inwardly directed nucleoside transport. The purpose of this study was to characterize the effects of adenosine on ion channels in A549 cells and the role of nucleoside transporters in this regulation. Ion replacement and pharmacological studies showed that adenosine and an inhibitor of human equilibrative nucleoside transporter (hENT)-1, nitrobenzylthioinosine, activated K(+) channels, most likely Ca(2+)-dependent intermediate-conductance K(+) (I(K)) channels. A(1) but not A(2) receptor antagonists blocked the effects of adenosine. RT-PCR studies showed that A549 cells expressed mRNA for I(K)-1 channels as well as A(1), A(2A), and A(2B) but not A(3) receptors. Similarly, mRNA for equilibrative (hENT1 and hENT2) but not concentrative (hCNT1, hCNT2, and hCNT3) nucleoside transporters was detected, a result confirmed in functional uptake studies. These studies showed that adenosine controls the function of K(+) channels in A549 cells and that hENTs play a crucial role in this process.