Merck
  • Influence of potassium fluoride on the syntheses of methylpiperazine-modified poly(vinyl chloride)s for use as fixed-site proton carrier membranes.
Influence of potassium fluoride on the syntheses of methylpiperazine-modified poly(vinyl chloride)s for use as fixed-site proton carrier membranes.
Journal of biomedical materials research (1998-03-10)
A R Roudman, R P Kusy
ABSTRACT
Aminated poly(vinyl chloride) (PVC) membranes were prepared that had a Nernstian response over a wide range of pH. The reaction between PVC and methyl-piperazine (MePIP) in dimethylformamide (DMF) was studied over a wide range of time and temperature, and in the presence of the catalyst, potassium fluoride (KF). Time, temperature, and KF increased the nitrogen (N) content of the resulting polymers, but sometimes at the expense of decreasing their specific viscosities (molecular weights). Activation energies of processes that occurred in different temperature ranges were estimated assuming an Arrhenius relationship. A Nernstian response occurred once the N content approached to about 0.3 w/w %, and was accelerated by the presence of KF at elevated temperatures. Increasing the N content above about 3% led to a loss of the Nernstian response either because of an increase in the double bond content and a subsequent decrease in polymer mobility, or because of a decrease in the molecular weight of the copolymer and concomitant difficulties in film preparation.
Sigma-Aldrich
1-Methylpiperazine, 99%
Supelco
1-Methylpiperazine, analytical standard