Merck
  • Direct identification of the common cheese contaminant Penicillium commune in factory air samples as an aid to factory hygiene.
Direct identification of the common cheese contaminant Penicillium commune in factory air samples as an aid to factory hygiene.
Letters in applied microbiology (1996-05-01)
F Lund
ABSTRACT
Creatine sucrose dichloran agar (CREAD) was used as a selective medium for Penicillium commune and related species found in air samples in a cheese factory. Using growth and simple colony characters on CREAD together with detection of indole metabolites with a filter paper method, it was possible to identify all 22 P. commune isolates from a total of 43 Penicillium isolates. Penicillium commune numbers on CREAD were compared with those found on a general isolation medium, dichloran 18% glycerol agar.
Sigma-Aldrich
2,6-Dichloro-4-nitroaniline, 96%