Merck
  • Interaction of N-methyl-anthraniloyl-labelled porcine apolipoprotein A-I with porcine lecithin: cholesterol acyltransferase: an energy transfer study.
Interaction of N-methyl-anthraniloyl-labelled porcine apolipoprotein A-I with porcine lecithin: cholesterol acyltransferase: an energy transfer study.
The International journal of biochemistry (1993-04-01)
G Knipping, A Hermetter, M R Hubmann
ABSTRACT
1. To investigate whether a direct protein-protein interaction between apoA-I and lecithin:cholesterol acyltransferase (LCAT) is necessary for the activation of the enzyme, apoA-I was labelled with N-methylisatoic anhydride at lysine residues. The intermolecular resonance energy transfer from tryptophan residues of LCAT (donor) to N-methyl-anthraniloyl (NMA)-labelled apoA-I (NMA-apoA-I) (acceptor) was used as a sensitive fluorescence method for studying molecular interactions. 2. In the absence of lipids no fluorescence energy transfer was measurable. 3. Fluorescence energy transfer occurred from LCAT to NMA-apoA-I in the presence of liposomes with phospholipid/cholesterol ratios ranging from 5:1 to 18:1 and regardless whether only 1 or up to 5 NMA-apoA-I molecules resided at the liposome surface. 4. This indicates a preferred binding of the enzyme directly to or in spatial proximity to the activator protein NMA-apoA-I even if enough space at the liposome surface is available to allow LCAT binding at a distance, where no energy transfer is measurable.
Sigma-Aldrich
N-Methylisatoic anhydride, technical, 90%