Merck
  • [Toxicologic and hygienic evaluation of benzyl acetate as an atmospheric air pollutant].
[Toxicologic and hygienic evaluation of benzyl acetate as an atmospheric air pollutant].
Gigiena i sanitariia (1993-10-01)
L A Tepikina, N P Zinovéva, Z V Shipulina
ABSTRACT
LD50 of benzyl-acetate (BA) was determined on the level 2500 mg/kg. Sensitivity of developed method for BA analysis--0.008-0.01 mg/m3. Single MAC of BA in environmental air is 0.01 mg/m3 (4th class of danger).
Product Description
Supelco
Benzyl acetate, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl acetate, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG