Merck
  • Palladium(II), magnesium(II), and barium(II) nitrate combinations for matrix modification in electrothermal atomic absorption measurement of total selenium in human urine.
Palladium(II), magnesium(II), and barium(II) nitrate combinations for matrix modification in electrothermal atomic absorption measurement of total selenium in human urine.
Analytical biochemistry (1994-08-01)
E N Drake, T D Hain
ABSTRACT
Electrothermal atomic absorption spectrometry with a temperature-stabilizing platform and palladium(II) nitrate-magnesium nitrate-barium nitrate matrix modifier combination was used to measure total selenium concentrations in 0.5-ml samples of human urine. The method was validated by analysis of desiccated, standard urine samples (SRM 2670) from NIST. The peak height response was linear from 0 to 150 micrograms Se/liter urine with a sensitivity of 0.001 absorbance unit/micrograms Se/liter and a detection limit of 6 micrograms Se/liter (P = 0.05) for a single measurement. Total urinary selenium concentrations and urine volumes of 24-h samples from 28 healthy adult subjects on self-selected diets and living in San Angelo, Texas were measured. Daily total urinary excretions of 16 male and 12 female subjects (mean +/- SD) were 28 +/- 26 and 34 +/- 25 micrograms Se/day, respectively. A single subject taking 300 micrograms Se/day in the form of aqueous SeO2 as a dietary supplement excreted 232 micrograms Se/day. The unsupplemented subjects' daily selenium excretions correspond to a mean dietary intake of approximately 60 micrograms/day.
Sigma-Aldrich
Barium nitrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium nitrate, 99.999% trace metals basis