Merck
  • Characterization of P-S bond hydrolysis in organophosphorothioate pesticides by organophosphorus hydrolase.
Characterization of P-S bond hydrolysis in organophosphorothioate pesticides by organophosphorus hydrolase.
Archives of biochemistry and biophysics (1995-04-01)
K Lai, N J Stolowich, J R Wild
ABSTRACT
The extensive use of organophosphorothioate insecticides in agriculture has resulted in the risk of environmental contamination with a variety of broadly based neurotoxins that inhibit the acetylcholinesterases of many different animal species. Organophosphorus hydrolase (OPH, EC 3.1.8.1) is a broad-spectrum phosphotriesterase that is capable of detoxifying a variety of organophosphorus neurotoxins by hydrolyzing various phosphorus-ester bonds (P-O, P-F, P-CN, and P-S) between the phosphorus center and an electrophilic leaving group. OPH is capable of hydrolyzing the P-X bond of various organophosphorus compounds at quite different catalytic rates: P-O bonds (kcat = 67-5000 s-1), P-F bonds (kcat = 0.01-500 s-1), and P-S bonds (kcat = 0.0067 to 167 s-1). P-S bond cleavage was readily demonstrated and characterized in these studies by quantifying the released free thiol groups using 5,5'-dithio-bis-2-nitrobenzoic acid or by monitoring an upfield shift of approximately 31 ppm by 31P NMR. A decrease in the toxicity of hydrolyzed products was demonstrated by directly quantifying the loss of inhibition of acetylcholinesterase activity. Phosphorothiolate esters, such as demeton-S, provided noncompetitive inhibition for paraoxon (a P-O triester) hydrolysis, suggesting that the binding of these two different classes of substrates was not identical.
Product Description
Supelco
Phosalone, PESTANAL®, analytical standard