Merck
  • Solid-phase synthesis of oxytocin using iodotrichlorosilane as Boc deprotecting agent.
Solid-phase synthesis of oxytocin using iodotrichlorosilane as Boc deprotecting agent.
International journal of peptide and protein research (1995-04-01)
K M Sivanandaiah, V V Sureshbabu, S C Shankaramma
ABSTRACT
Deprotection of the tert-butoxycarbonyl group during solid-phase synthesis of peptides can be conveniently and efficiently carried out using a neutral reagent, silicon tetrachloride/sodium iodide (iodotrichlorosilane). This simple and rapid method has been advantageously employed during the solid-phase synthesis of the pituitary hormone, oxytocin.
Sigma-Aldrich
Silicon tetrachloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Silicon tetrachloride, 99%
Sigma-Aldrich
Silicon tetrachloride, 99.998% trace metals basis
Sigma-Aldrich
Silicon tetrachloride, packaged for use in deposition systems