Merck
  • Quantification of cells in islets of Langerhans using DNA determination.
Quantification of cells in islets of Langerhans using DNA determination.
Acta diabetologica latina (1981-01-01)
J Beckmann, S Holze, S Lenzen, U Panten
ABSTRACT
DNA content seems to be an ideal reference parameter for data on secretory function or metabolism of pancreatic islets. The approved fluorometric DNA assay with diaminobenzoic acid (DABA) of Kissane and Robins comprises repeated ethanol extractions of the tissue for removal of lipids from which some DABA-reactive aldehydes may originate. In the present study it is demonstrated that only negligible amounts of DABA-positive material are extractable from islets of Langerhans. Furthermore, it is shown that various substances used in experiments on the endocrine pancreas do not interfere with the DABA-DNA reaction. A modification of the original DABA procedure which does not include ethanol extractions and which is thus more simple and accurate is described for application to pancreatic islets in the absence as well as in the presence of incubation medium. A close linear correlation between islet dry weight and islet DNA content is demonstrated. Islets from rats, normal mice, and ob/ob mice contain 38.3-39.2 ng DNA per microgram dry weight.
Sigma-Aldrich
3,5-Diaminobenzoic acid, 98%