Merck
  • The distribution of euchrysine-binding grains of the peripheral and bone marrow stem cells in different types of acute leukemia.
The distribution of euchrysine-binding grains of the peripheral and bone marrow stem cells in different types of acute leukemia.
Archivum immunologiae et therapiae experimentalis (1984-01-01)
B Mdzewski, Z Tomaszewska, B Romer, R Rokicka, J Krawczyński, A Kendziorek, D Derulska
ABSTRACT
The distribution of the fluorescent euchrysine-binding grains in the peripheral and bonmarrow stem cells was estimated in 28 cases of different types of acute leukemia verified according to Lòffler's cytochemical classification. The relationship between the pattern of fluorescent euchrysine-binding grains of human stem cells and the cytochemical type of acute leukemias has been established. Consequently, the authors suggest that the euchrysine-binding grains pattern of human stem cells may have a diagnostic value in distinguishing between various types of acute leukemia.
Sigma-Aldrich
Acridine Orange hemi(zinc chloride) salt, Dye content 85 %
Pricing and availability is not currently available.
Sigma-Aldrich
Acridine Orange hemi(zinc chloride) salt, For nucleic acid staining in cells or gels
Pricing and availability is not currently available.