Merck
  • Mutagenicity tests of lipid oxidation products in Salmonella typhimurium: monohydroperoxides and secondary oxidation products of methyl linoleate and methyl linolenate.
Mutagenicity tests of lipid oxidation products in Salmonella typhimurium: monohydroperoxides and secondary oxidation products of methyl linoleate and methyl linolenate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1985-12-01)
J T MacGregor, R E Wilson, W E Neff, E N Frankel
ABSTRACT
Nine hydroperoxy and hydroperoxy-epidioxy oxidation products derived from either autoxidation (AO) or photosensitized oxidation (PO) of methyl linoleate (MLo) or methyl linolenate (MLn) were tested for mutagenic activity by the Salmonella typhimurium his+ reversion assay using strains TA100, TA98, TA102, TA97 and TA1537. All nine oxidation products, monohydroperoxides from AO-MLn (I) or from PO-MLn (II), dihydroperoxides from PO-MLo (III), AO-MLn (IV) or PO-MLn (V), hydroperoxy epidioxides from PO-MLo (VI), AO-MLn (VII) or PO-MLn (VIII) and hydroperoxy bis-epidioxides from PO-MLn (IX), were weakly mutagenic in strains TA97 and/or TA100. The hydroperoxy epidioxides (VI-IX) exhibited significantly greater activity in strain TA97 than did the monohydroperoxides (I, II) or the dihydroperoxides (III-V). In strain TA100, all of the oxidation products tested exhibited similar activity. No major differences between products derived from autoxidized and photooxidized MLn (I v. II, IV v. V, VII v. VIII) were obtained. Rat-liver S-9 reduced the toxicity of all oxidation products to the tester strains. The greatest mutant yields were usually obtained in the presence of S-9, but mutagenic potency was sometimes greater without S-9. The structural feature common to all of the mutagenic oxidation products was the presence of a hydroperoxy group, suggesting that this characteristic is responsible for the observed mutagenicity, either directly or through a common degradative pathway to reactive products of lower molecular weight.
Sigma-Aldrich
Methyl linolenate, technical, 70-80% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl linolenate, ≥99% (GC)
Pricing and availability is not currently available.
Supelco
Methyl linolenate, analytical standard
Pricing and availability is not currently available.