Merck
Poly(hydroxy acids) in drug delivery.
Critical reviews in therapeutic drug carrier systems (1987-01-01)
K Juni, M Nakano
ABSTRACT
Poly(hydroxy acids) so far have been examined for use in drug delivery in limited number, while the advantageous use of the polymers has been recognized due to their biodegradability and biocompatibility. Homo- and copolymers of lactic acid and glycolic acid have been studied in drug delivery by many workers, while homo- and copolymers of epsilon-caprolactone have been studied by only one group of workers. Although poly-hydroxybutyric acid had been found to be a naturally occurring polymer, examination as to the use of the polymer in drug delivery is rather recent and reports are still limited. In the present article, the use of poly(hydroxy acids) including homo- and copolymers of lactic acid and glycolic acid, polycaprolactone, and poly-beta-hydroxybutyric acid in drug delivery is reviewed. Physicochemical properties, biodegradability, and biocompatibility of the polymers, and evaluations in vitro and in vivo of specific dosage forms using the polymers, are included. The most recent work in our laboratories on the use of polyactic acid and poly-beta-hydroxybutyric acid is also included.
Sigma-Aldrich
Poly(3-hydroxybutyric acid), average Mn 10,000
Sigma-Aldrich
Poly((R)-3-hydroxybutyrate-co-(R)-3-hydroxyvalerate-co-(R)-3-hydroxyhexanoate), natural origin, PHH content 4.1 %, PHV content 2.3 %
Sigma-Aldrich
Poly(3-hydroxybutyric acid), natural origin, average Mn ~500,000
Sigma-Aldrich
ε-Decalactone, ≥99%
Sigma-Aldrich
Poly((R)-3-hydroxybutyrate-co-(R)-3-hydroxyhexanoate), natural origin, PHH content 15.2 %