Merck
  • Strongly enhanced toxicity of the mushroom toxin alpha-amanitin by an amatoxin-specific Fab or monoclonal antibody.
Strongly enhanced toxicity of the mushroom toxin alpha-amanitin by an amatoxin-specific Fab or monoclonal antibody.
Toxicon : official journal of the International Society on Toxinology (1988-01-01)
H Faulstich, K Kirchner, M Derenzini
ABSTRACT
A monoclonal antibody, with high affinity against the mushroom toxin alpha-amanitin, was prepared. Administration of the Fab fragment of the monoclonal antibody to mice caused a 50-fold increase in alpha-amanitin toxicity. Electron micrographs showed normal appearance of hepatocytes but typical, amanitin-induced lesions in cells of the proximal convoluted tubules of the kidney. The pronounced nephrotoxicity is mainly explained by glomerular filtration and tubular reabsorption of the Fab-amatoxin complex and, to a lesser extent, of the immunoglobulin-amatoxin complex, which is still c. Twice as toxic as free alpha-amanitin. To our knowledge this is the first reported case where immunoglobulins or their fragments enhance rather than decrease the activity of a toxin. Accordingly, immunotherapy of Amanita mushroom poisoning in humans does not appear promising.
Sigma-Aldrich
β-Amanitin from Amanita phalloides, ≥85% (HPLC)