Merck
  • Terpenoid biotransformation in mammals. V. Metabolism of (+)-citronellal, (+-)-7-hydroxycitronellal, citral, (-)-perillaldehyde, (-)-myrtenal, cuminaldehyde, thujone, and (+-)-carvone in rabbits.
Terpenoid biotransformation in mammals. V. Metabolism of (+)-citronellal, (+-)-7-hydroxycitronellal, citral, (-)-perillaldehyde, (-)-myrtenal, cuminaldehyde, thujone, and (+-)-carvone in rabbits.
Xenobiotica; the fate of foreign compounds in biological systems (1989-08-01)
T Ishida, M Toyota, Y Asakawa
ABSTRACT
1. The metabolism of (+)-citronellal, (+-)-7-hydroxycitronellal, citral, (-)-perillaldehyde, (-)-myrtenal, cuminaldehyde, thujone, and (+-)-carvone was studied in rabbits. 2. In (+-)-7-hydroxycitronellal, (-)-perillaldehyde, (-)-myrtenal and cuminaldehydes, both primary alcohols and carboxylic acids were formed. 3. In (-)-citronellal and citral, regioselective oxidation was found and a trans-positioned methyl group was carboxylated in each case. 4. In o-cuminaldehyde, reduction but not oxidation, was found.
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG
Sigma-Aldrich
Cuminaldehyde, 98%
Sigma-Aldrich
Cuminaldehyde, ≥98%, FCC, FG
Sigma-Aldrich
7-Hydroxycitronellal, Odorant used in allergy studies