Merck
  • Performance study and kinetic modeling of hybrid bioreactor for treatment of bi-substrate mixture of phenol-m-cresol in wastewater: process optimization with response surface methodology.
Performance study and kinetic modeling of hybrid bioreactor for treatment of bi-substrate mixture of phenol-m-cresol in wastewater: process optimization with response surface methodology.
Journal of environmental sciences (China) (2013-08-09)
Sudipta Dey, Somnath Mukherjee
ABSTRACT
Performance of a hybrid reactor comprising of trickling filter (TF) and aeration tank (AT) unit was studied for biological treatment of wastewater containing mixture of phenol and m-cresol, using mixed microbial culture. The reactor was operated with hydraulic loading rates (HLR) and phenolics loading rates (PLR) between 0.222-1.078 m3/(m2 x day) and 0.900-3.456 kg/(m3 x day), respectively. The efficiency of substrate removal varied between 71%-100% for the range of HLR and PLR studied. The fixed film unit showed better substrate removal efficiency than the aeration tank and was more resistant to substrate inhibition. The kinetic parameters related to both units of the reactor were evaluated and their variation with HLR and PLR were monitored. It revealed the presence of substrate inhibition at high PLR both in TF and AT unit. The biofilm model established the substrate concentration profile within the film by solving differential equation of substrate mass transfer using boundary problem solver tool 'bvp4c' of MATLAB 7.1 software. Response surface methodology was used to design and optimize the biodegradation process using Design Expert 8 software, where phenol and m-cresol concentrations, residence time were chosen as input variables and percentage of removal was the response. The design of experiment showed that a quadratic model could be fitted best for the present experimental study. Significant interaction of the residence time with the substrate concentrations was observed. The optimized condition for operating the reactor as predicted by the model was 230 mg/L of phenol, 190 mg/L of m-cresol with residence time of 24.82 hr to achieve 99.92% substrate removal.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology