Merck
  • A technique to create an interocclusal bite registration using in situ implant healing abutments.
A technique to create an interocclusal bite registration using in situ implant healing abutments.
The Journal of the Tennessee Dental Association (2013-08-06)
Russell Wicks, Swati Ahuja, Vinay Jain, Cimara Fortes Ferreira
ABSTRACT
Obtaining an accurate interocclusal record which relates the upper and lower jaws to one another is critical in the laboratory production of dental restorations. This may be particularly challenging when the configuration of missing teeth dictate that such records are dependent on the remaining mucosal tissues for support. This article presents a practical and time saving method of using implant healing abutments, located within an edentulous distal extension space, to serve as vertical stops for an interocclusal record.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic