Merck
  • Photocatalytic performance of nitrogen, osmium co-doped TiO2 for removal of eosin yellow in water under simulated solar radiation.
Photocatalytic performance of nitrogen, osmium co-doped TiO2 for removal of eosin yellow in water under simulated solar radiation.
Journal of nanoscience and nanotechnology (2013-08-02)
Alex T Kuvarega, Rui W M Krause, Bhekie B Mamba
ABSTRACT
Nitrogen, osmium co-doped TiO2 photocatalysts were prepared by a modified sol-gel method using ammonia as the nitrogen source and osmium tetroxide as the source of osmium. The role of rutile phase OsO2 in enhancing the photocatalytic activity of rutile TiO2 towards the degradation of Eosin Yellow was investigated. The materials were characterised by various techniques that include FTIR, Raman, XRD, SEM, EDS, TEM, TGA and DRUV-Vis. The amorphous, oven dried sample was transformed to the anatase and then the rutile phase with increasing calcination temperature. DRUV-Vis analysis revealed a red shift in absorption with increasing calcination temperature, confirmed by a decrease in the band gap of the material. The photocatalytic activity of N, Os co-doped TiO2 was evaluated using eosin yellow degradation and activity increased with increase in calcination temperature under simulated solar irradiation. The rutile phase of the co-doped TiO2 was found to be more effective in degrading the dye (k(a) = 1.84 x 10(-2) min(-1)) compared to the anatase co-doped phase (k(a) = 9.90 x 10(-3) min(-1)). The enhanced photocatalytic activity was ascribed to the synergistic effects of rutile TiO2 and rutile OsO2 in the N, Os co-doped TiO2.
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Osmium, powder, 99.9% trace metals basis
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Titanium, IRMM®, certified reference material, 0.5 mm foil
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Eosin Y, 75% (HPLC)