Merck
Preparation of ZnO nanowire using sludge from wastewater treatment.
Journal of nanoscience and nanotechnology (2013-07-26)
Xiaolong Li, Fengqin Zhang, Chao Ma, Sauli Elingarami, Nongyue He
ABSTRACT
In this study, we present the synthesis of ZnO nanowire by hydrothermal process through reutilization of sludge from soy sauce wastewater electrochemical treatment. The influences of floc content and caramel pigment concentration on the morphologies of ZnO were studied. The products were characterized by XRD and SEM. The results showed that ZnO powder was of hexagonal wurtzite structure and well crystallized with high purity. Floc content and caramel pigment concentration played an important role in the final morphologies of ZnO nanowires. On the basis of these results, a possible growth mechanism for the formation of ZnO nanowires was described. These findings not only may contribute to control the synthesis of ZnO, but also present a novel method for the disposal of wastewater treatment sludge.
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%