Merck
  • Effect of particle size of PtRu nanoparticles embedded in WO3 on electrocatalysis.
Effect of particle size of PtRu nanoparticles embedded in WO3 on electrocatalysis.
Journal of nanoscience and nanotechnology (2013-07-19)
Sung Jong Yoo, Hee Young Park, Seung Jun Hwang, Sung Gyu Pyo, Soo-Kil Kim, Yung-Eun Sung, Tae-Hoon Lim
ABSTRACT
Size-controlled PtRu nanoparticles embedded in WO3 were prepared by simultaneous multigun sputtering on pure targets of Pt, Ru, and WO3. The mean diameter of the PtRu nanoparticles, as confirmed by high-resolution transmission electron microscopy, can be varied from -2.3 to -3.6 nm by varying the RF power ratio of PtRu and WO3. On the basis of transmission electron diffraction results for the PtRu nanoparticles embedded in WO3, it was confirmed that PtRu exists as an alloy metal phase, whereas the WO3 matrix is present as an amorphous phase. Size-controlled PtRu/WO3 electrodes were found to exhibit unique electronic properties depending on their size, which affected the potential of zero total charge and the methanol oxidation reaction. The mass activity of PtRu/WO3 for methanol oxidation was determined by the interplay of the surface electronic factors at the metal-solution interface; the oxophilicity of the nanoparticles increased with decreasing particle size.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%