Merck
  • [Analysis of factors affecting the duration of treatment with sorafenib in patients with hepatocellular carcinoma].
[Analysis of factors affecting the duration of treatment with sorafenib in patients with hepatocellular carcinoma].
Gan to kagaku ryoho. Cancer & chemotherapy (2013-07-16)
Noriaki Kitada, Kengo Kanamori, Ayako Konishi, Shoji Tanaka, Yoshiki Suginoshita, Tetsuro Inokuma, Tohru Hashida
ABSTRACT
The current status of treatment with sorafenib, and factors affecting the duration of treatment in patients started on sorafenib for hepatocellular carcinoma from July 2009 until April 2011 in the Department of Gastroenterology at Kobe City Medical Center General Hospital, were examined. Of 21 patients, 12 were able to continue the administration of sorafenib for more than one month, but 9 had to be discontinued within one month due to disease progression, worsening of general condition, and severe adverse reactions. In the group that was discontinued early, the rate of discontinuation due to side effects such as general fatigue, diarrhea, and hepatic encephalopathy was higher than in the long-term treatment group. On the other hand, hand-foot syndrome developed only in one case in both groups. The median value of PIVKA- II at the start of treatment in the long-term and early discontinued treatment groups were 672.5 and 14, 203 mAU/mL, respectively, and the values in the long-term group were significantly lower than those in the early-discontinued group (p < 0.05). From these results, the values of PIVKA-II at the start of sorafenib were considered to be factors affecting the continuation of sorafenib treatment. In addition, the dosing period was considered to be extended to focus on measures to take against the side effects of sorafenib within the early phase. Therefore, it was considered that these factors improved the effect of treatment with sorafenib in patients with hepatocellular carcinoma.
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture