Merck
Modeling of crystal violet adsorption by bottom ash column.
Water environment research : a research publication of the Water Environment Federation (2013-07-10)
P V Nidheesh, R Gandhimathi, S T Ramesh, T S Anantha Singh
ABSTRACT
The removal of crystal violet from wastewater, by means of bottom ash, was investigated in a packed bed down-flow column. The bed depth service time (BDST) model was used to analyze the experimental data up to breakthrough time (corresponding to C(t)/C(0) = 0.1). A mass transfer model was used to analyze the mass transfer zone. The breakthrough curve was analyzed by the Thomas, Yoon-Nelson, and Clark models. All models fit well with the experimental data. Results showed that as the flow rate increases, at a constant concentration and bed depth, the value of the adsorption capacity of bottom ash decreases. The adsorption capacity of bottom ash decreases with an increase in depth and initial crystal violet concentration. Error analyses were performed for the Thomas, Yoon-Nelson, and Clark models. Water
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution