Merck
  • Formulation development and evaluation of Diltiazem HCl sustained release matrix tablets using HPMC K4M and K100M.
Formulation development and evaluation of Diltiazem HCl sustained release matrix tablets using HPMC K4M and K100M.
Pakistan journal of pharmaceutical sciences (2013-07-03)
Faaiza Qazi, Muhammad Harris Shoaib, Rabia Ismail Yousuf, Tanveer Mustafa Qazi, Zafar Alam Mehmood, S M Farid Hasan
ABSTRACT
The aim of this study was to develop a sustained release hydrophilic matrix tablet of Diltiazem HCl and evaluates the effect of formulation variables (e.g. lubricant, binder, polymer content and viscosity grades of HPMC) on drug release. Twelve different formulations (F1-F12) were prepared by direct compression. The results of the physical parameters and assay were found to be within the acceptable range. Rate of drug release was found to be slow as the fraction of the polymer was increased from 20-50%. The drug release rate from tablets containing K4M was effectively controlled by increasing the talc concentration, whereas the burst effect was reduced by increasing binder content. The drug release was higher with K4M as compare to K100M. Model-dependent and independent methods were used for data analysis and the best results were observed for K4M in Higuchi (R(2)=0.9903-0.9962) and K100M in Baker and Lonsdale (R(2)=0.9779-0.9941). The release mechanism of all formulations was non-Fickian. F7 (50% K4M, 2% talc, 10% Avicel PH101) and F11 (40% K100M) were very close to targeted release profile. F12 (50% K100M) exhibited highest degree of swelling and lowest erosion. The f1 and f2 test were performed taking F11 as a reference formulation.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard