Merck
  • Biochemical parameters of Saccharopolyspora erythraea during feeding ammonium sulphate in erythromycin biosynthesis phase.
Biochemical parameters of Saccharopolyspora erythraea during feeding ammonium sulphate in erythromycin biosynthesis phase.
Prikladnaia biokhimiia i mikrobiologiia (2013-06-26)
X Zou, W-J Li, W Zeng, H-F Hang, J Chu, Y P Zhuang, S L Zhang
ABSTRACT
The physiology of feeding ammonium sulphate in erythromycin biosynthesis phase of Saccharopolyspora erythraea on the regulation of erythromycin A (Er-A) biosynthesis was investigated in 50 L fermenter. At an optimal feeding ammonium sulphate rate of 0.03 g/L per h, the maximal Er-A production was 8281 U/mL at 174 h of growth, which was increased by 26.3% in comparison with the control (6557 U/mL at 173 h). Changes in cell metabolic response of actinomycete were observed, i.e. there was a drastic increase in the level of carbon dioxide evolution rate and oxygen consumption. Assays of the key enzyme activities and organic acids of S. erythraea and amino acids in culture broth revealed that cell metabolism was enhanced by ammonium assimilation, which might depend on the glutamate transamination pathway. The enhancement of cell metabolism induced an increase of the pool of TCA cycle and the metabolic flux of erythromycin biosynthesis. In general, ammonium assimilation in the erythromycin biosynthesis phase of S. erythraea exerted a significant impact on the carbon metabolism and formation of precursors of the process for dramatic regulation of secondary metabolites biosynthesis.
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
L-Glutamic acid hydrochloride
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%