Merck
  • Corneal collagen cross-linking in the stabilization of PRK, LASIK, thermal keratoplasty, and orthokeratology.
Corneal collagen cross-linking in the stabilization of PRK, LASIK, thermal keratoplasty, and orthokeratology.
Current opinion in ophthalmology (2013-06-06)
Michelle K Nguyen, Roy S Chuck
ABSTRACT
To describe the use of corneal collagen cross-linking (CXL) and its efficacy in the stabilization of keratorefractive procedures, including PRK, laser in-situ keratomileusis (LASIK), thermal keratoplasty, and orthokeratology. Since its introduction, CXL has quickly gained interest in the treatment of ectactic disorders due to its ability to increase the biomechanical stability of the cornea. In its earliest use, it has shown to be effective in the treatment of both keratoconus and post-LASIK ectasia. More recent studies of CXL in combination with keratorefractive procedures have shown varying degrees of success. CXL with PRK has shown to be effective in slowing or halting the progression of keratoconus, pellucid marginal degeneration, and post-LASIK ectasia, in addition to potentially decreasing or delaying the need for penetrating keratoplasty. Several small case series have also demonstrated improved stability and efficacy of PRK and LASIK when combined with CXL, as well as a potentially decreased risk of postprocedure ectasia. In conjunction with thermokeratoplasty and orthokeratology, CXL has shown improved but only temporary results in the treatment of keratoconus. Future studies are needed to determine the efficacy and long-term stability of CXL in combination with keratorefractive procedures, as well as to address possible complications.
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance