Merck
  • Spectrometric method for the assay of ramipril using molybdophosphoric acid.
Spectrometric method for the assay of ramipril using molybdophosphoric acid.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (2013-05-25)
Mădălina Vieriu, Nela Bibire, Gladiola Tântaru, M Apostu, Alina Diana Panainte, V Dorneanu
ABSTRACT
Ramipril is a drug of the angiotensin converting enzyme inhibitor class. A new molecular absorbance spectrometric method was developed for the assay of ramipril using molybdophosphoric acid in acidic medium. The reaction product showed a maximum absorbance at 361 nm. The optimum conditions of the reaction were established. The developed method was validated. The method showed a good linearity in the range of 8 - 36 microg/ml (correlation coefficient r = 0.9996). The detection limit (LD) was 0.86microg/ml and the quantification limit (LQ) 2.88 pg/ml. Precision and accuracy were determined (RSD = 1.15%); mean recovery was 98.90% in the 97.13-101.03% concentration range. The proposed method is simple, easy to perform, sensitive, linear, precise, accurate and robust.
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium phosphomolybdate hydrate, technical
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent, for chromatography
Sigma-Aldrich
Ramipril, ≥98% (HPLC)
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis