Merck
  • Repeated pulse intramuscular injection of pralidoxime chloride in severe acute organophosphorus pesticide poisoning.
Repeated pulse intramuscular injection of pralidoxime chloride in severe acute organophosphorus pesticide poisoning.
The American journal of emergency medicine (2013-05-18)
Xue Tang, Ruilan Wang, Hui Xie, Jiachang Hu, Wenbiao Zhao
ABSTRACT
This study aimed to clarify the efficacy of 2 therapies for patients with severe acute organophosphorus pesticide poisoning, including atropine adverse effects, the length of intensive care unit (ICU) stay, complications, and mortality. A retrospective cohort study of 152 cases collected from May 2008 to November 2012 at 2 urban university hospitals was conducted. Patients admitted to the hospital for organophosphate poisoning were divided into 2 groups with different therapeutic regimens: group A was administered a repeated pulse intramuscular injection of pralidoxime chloride, and group B received the same initial dosage of atropine and pralidoxime chloride, but pralidoxime chloride intravenous therapy was administered for only 3 days, regardless of the length of atropine therapy. Subsequently, atropine adverse effects, length of ICU stay, complications, and mortality were statistically analyzed and compared between the 2 groups. The total dose of atropine was 57.40 ± 15.14 mg in group A and 308.26 ± 139.16 mg in group B; group A received less atropine than did group B (P = .001). The length of ICU stay in group A was reduced (P = .025), and group A had fewer atropine adverse effects (P = .002). However, there was no significant difference in the mortality or complication rate between the 2 groups (P > .05). In patients with severe poisoning, group A used less atropine, had fewer atropine adverse effects, and had a shorter ICU stay. We suggest that therapy should be started as early as possible using a sufficient amount of pralidoxime chloride started intramuscularly in combination with atropine and that the drugs should not be prematurely discontinued.
Sigma-Aldrich
2-Pyridinealdoxime methiodide, 99%
Sigma-Aldrich
Pyridine-2-aldoxime methochloride