Merck
  • Seven clinical conundrums in the treatment of ANCA-associated vasculitis.
Seven clinical conundrums in the treatment of ANCA-associated vasculitis.
Clinical and experimental rheumatology (2013-07-24)
M A Alba, L F Flores-Suárez
ABSTRACT
Granulomatosis with polyangiitis and microscopic polyangiitis are two autoimmune diseases characterised by necrotising small-vessel vasculitis and presence of antineutrophil cytoplasm autoantibodies (ANCA). Current immunosuppressive regimes that combine cyclophosphamide and glucocorticoids have dramatically improved the outcome for these patients. However, these treatments are associated with toxic effects and do not lead to permanent remission in the majority of cases. Newer approaches have been sought during the last 15 years, with improvement in medication protocols and inclusion of novel therapies. This review develops on seven clinical conundrums of evidence-based therapeutic strategies for ANCA-vasculitis, posed as questions on aspects such as the role of established drugs in both remission induction and maintenance: glucocorticoids (and its duration), oral cyclophosphamide, methotrexate, TNF-α blockers, plasma exchange, mycophenolate mofetil, plus one related to newer developments in treatment with agents blocking the complement system and the possible role of sequential or combined therapies, mainly directed against B cells.
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard