Merck
  • Effect of amiloride on experimental acid-induced heartburn in non-erosive reflux disease.
Effect of amiloride on experimental acid-induced heartburn in non-erosive reflux disease.
Digestive diseases and sciences (2013-05-08)
William J Bulsiewicz, Nicholas J Shaheen, Mark B Hansen, Amy Pruitt, Roy C Orlando
ABSTRACT
Acid-sensing ion channels (ASICs) are esophageal nociceptors that are candidates to mediate heartburn in non-erosive reflux disease (NERD). Amiloride, a diuretic, is known to inhibit ASICs. For this reason, we sought a role for ASICs in mediating heartburn by determining whether amiloride could block heartburn in NERD induced by esophageal acid perfusion. In a randomized double-blind crossover study, we perfused the esophagus with amiloride or (saline) placebo prior to eliciting acid-induced heartburn in patients with a history of proton pump inhibitor-responsive NERD. Those with NERD and positive modified Bernstein test were randomized to perfusion with amiloride, 1 mmol/l, or placebo for 5 min, followed by repeat acid-perfusion. Heartburn severity and time to onset was measured and the process repeated following crossover to the alternative agent. 14 subjects completed the study. Amiloride did not reduce the frequency (100 vs. 100 %) or severity of acid-induced heartburn (Mean 2.50 ± SEM 0.33 vs. 2.64 ± 0.45), respectively. There was a trend towards longer time to onset of heartburn for amiloride versus placebo (Mean 2.93 ± SEM 0.3 vs. 2.36 ± 0.29 min, respectively), though these differences did not reach statistical significance (p > 0.05). Amiloride had no significant effect on acid-induced heartburn frequency or severity in NERD, although there was a trend towards prolonged time to onset of symptoms.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, derivatization grade (GC derivatization), LiChropur