Merck
Aqueous phase reforming of glycerol over nanosize Cu-Ni catalysts.
Journal of nanoscience and nanotechnology (2013-05-08)
Ji Yeon Kim, Seong Hak Kim, Dong Ju Moon, Jong Ho Kim, Nam Cook Park, Young Chul Kim
ABSTRACT
In this work, hydrogen production from glycerol by aqueous phase reforming (APR) is studied by using nanosize Ni-Cu catalysts supported on LaAlO3 perovskite in order to investigate the effects of the copper loading amount and the reaction conditions. Nanosize copper-promoted nickel-based catalysts were prepared by the precipitation method. The structure of the nanosize catalysts is characterized by XRD analysis. The surface area, morphology, dispersion and reducibility of the nanosize catalysts is examined by BET, TEM and TPR, respectively. It was found that 15Ni-5Cu/LaAlO3 catalyst showed the highest glycerol conversion and hydrogen selectivity. The highest activity found in the 15Ni-5Cu/LaAlO3 was attributed to it having the proper copper loading amount. It also has the lowest metal crystal size and the highest surface area, which have an effect on the catalytic activity and hydrogen selectivity. The 15Ni-5Cu/LaAlO3 catalyst showed the best performance for hydrogen production at a reaction temperature of 250 degrees C, a reaction pressure of 20 bar and a feed rate of 5 ml/h.
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Calcium titanate, nanopowder, <100 nm particle size (BET), 99% trace metals basis
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%