Merck
  • [Induction of adventitious roots of Echinacea pallida and accumulation of caffeic acid derivatives].
[Induction of adventitious roots of Echinacea pallida and accumulation of caffeic acid derivatives].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-01)
Chun-Hua Wu, Tao Huang, Xi-Hua Cui, Keeyoeup Paek
ABSTRACT
To investigate the effect of auxins 2,4-D,IAA,IBA,NAA on induction of adventitious roots as well as that of IBA concentrations on the growth of adventitious roots and the accumulation of caffeic acid derivatives, with test-tube seedling leaves Echinacea pallida as the explant, and cultivate adventitious roots in bioreactors. 1.0 mg x L(-1) IBA was found the best for the induction of adventitious roots, with the numer of induced adventitious roots up to 22. 5 in each culture dish. Among different concentrations for suspension cultivation of IBA tested, 1.0 mg x L(-1) IBA was found the most suitable for the growth of adventitious roots and the accumulation of caffeic acid derivatives. In a 5 L balloon type bubble bioreactor, 8.98 g x L(-1) dry weight was achieved after one month, which was 2.05 times of 4.38 g x L(-1) dry weight cultivated in a triangular flask. The content of echinacoside cultivated in a bioreactor was 14.08 mg x g(-1) DW, which was 2.4 times of cultivated roots. The contents of chlorogenic acid, chicoric acid and total caffeic acid derivatives were 4.0-25.6 times of ultivated roots. The study can provide high-quality biomedical drugs containing such caffeic acid derivatives as echinacoside for mass production of Echinacea purpurea medicines.
Product Description
Supelco
2,4-D, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Supelco
2,4-D, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
Indole-3-butyric acid, ≥98.0% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
Indole-3-butyric acid, BioReagent, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
€542.00