Merck
[Chemical constituents from cell cultures of Morus alba].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-01)
Xiao-Yu Tao, De-Wu Zhang, Ri-Dao Chen, Yun-Ze Yin, Jian-Hua Zou, Dan Xie, Lin Yang, Chun-Mei Wang, Jun-Gui Dai
ABSTRACT
The column chromatography on silica gel, Sephadex LH-20 and semi-preparative HPLC were used to separate and purify the compounds from the EtOAc extract of medium and MeOH extract of cell cultures of Morus alba. Eight compounds were isolated. Based on physico-chemical properties and spectroscopic data, their structures were identified as isobavachalcone (1), genistein (2), norartocarpetin (3), albanin A (4), guangsangon E (5), mulberrofuran F (6), chalcomoracin (7), kuwanon J (8). Compounds 3-6 were isolated from the cell cultures of M. alba for the first time.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-75, Medium
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-25, Fine
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Sigma-Aldrich
Sephadex® G-100, BioReagent, for molecular biology, DNA grade, medium
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Sephadex® G-25, BioReagent, for molecular biology, DNA grade, Superfine
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000