Merck
Two new compounds from Schisandra glaucescens.
Journal of Asian natural products research (2013-04-26)
Pei-Pei Zhang, Sha-Sha Gao, Tian-Tian Zhang, Xiao-Gang Wang, Guo-Liang Qing, Hong-Liang Jiang, Jia-Chun Chen, Hong-Quan Duan, Jin-Bo Fang
ABSTRACT
One new lignan (7S,8R,7'R,8'R)-7-(3,4-methylenedioxyphenyl)-8,8'-dimethyl-8'-hydroxyl-7'-methoxyl-7'-(3',4'-methylenedioxyphenyl)-tetrahydrofuran (1), one new sesquiterpene 2-hydroxy-11,12-dehydrocalamenene (2), one new natural product erythro-1-(3,4-dimethoxyphenyl)-4-(3,4-methylenedioxyphenyl)-2,3-dimethyl-butane (3), and two known lignans (+)-anwulignan(erythro-1-(4-hydroxy-3-methoxyphenyl)-4-(3,4-methylenedioxyphenyl)-2,3-dimethyl-butane) (4) and ( - )-zuonin-A (5) were isolated from the stems of Schisandra glaucescens Diels. Their structures were elucidated by spectroscopic methods. The cytotoxicity of compounds 1 and 2 was assayed.
Product Description
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%