Merck
  • Treatment of congenitally missing maxillary lateral incisors: an interdisciplinary approach.
Treatment of congenitally missing maxillary lateral incisors: an interdisciplinary approach.
The Journal of oral implantology (2013-04-25)
Brian J Jackson, Mark R Slavin
ABSTRACT
Dentists have several treatment options when restoring patients with congenitally missing maxillary lateral incisors. Endosseous implant therapy combined with orthodontics can achieve predictable outcomes while meeting patient expectations. This case report exhibits an interdisciplinary approach involving orthodontics, implant surgery, and prosthodontics to resolve this common dental concern.
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zinc phosphate, 99.998% trace metals basis