Merck
  • Methylation-dependent and -independent genomic targeting principles of the MBD protein family.
Methylation-dependent and -independent genomic targeting principles of the MBD protein family.
Cell (2013-04-16)
Tuncay Baubec, Robert Ivánek, Florian Lienert, Dirk Schübeler
ABSTRACT
To gain insight into the cellular readout of DNA methylation, we established a strategy for systematically profiling the genome-wide distribution of chromatin-interacting factors. This enabled us to create genomic maps for the methyl-CpG-binding domain (MBD) family of proteins, including disease-relevant mutants, deletions, and isoforms. In vivo binding of MBD proteins occurs predominantly as a linear function of local methylation density, requiring functional MBD domains and methyl-CPGs. This interaction directs specificity of MBD proteins to methylated, CpG-dense, and inactive regulatory regions. In contrast, binding to unmethylated sites varies between MBD proteins and is mediated via alternative domains or protein-protein interactions. Such targeting is exemplified by NuRD-complex-mediated tethering of MBD2 to a subset of unmethylated, active regulatory regions. Interestingly, MBD3 also occupies these sites, but like MBD2, binding is independent of the presence of hydroxymethylation. These functional binding maps reveal methylation-dependent and -independent binding modes and revise current models of DNA methylation readout through MBD proteins.
Product Description
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, meets USP testing specifications