Merck
  • Tandem ring-closing metathesis/isomerization reactions for the total synthesis of violacein.
Tandem ring-closing metathesis/isomerization reactions for the total synthesis of violacein.
Organic letters (2013-04-05)
Mette T Petersen, Thomas E Nielsen
ABSTRACT
A series of 5-substituted 2-pyrrolidinones was synthesized through a one-pot ruthenium alkylidene-catalyzed tandem RCM/isomerization/nucleophilic addition sequence. The intermediates resulting from RCM/isomerization showed reactivity toward electrophiles in aldol condensation reactions which provided a new entry for the total synthesis of the antileukemic natural product violacein.
Sigma-Aldrich
Violacein from Janthinobacterium lividum, >98% (violacein (minimum 85% violacein) and deoxyviolacein, HPLC)
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis