Merck
  • Ammonia-induced cell envelope injury in Escherichia coli and Enterobacter aerogenes.
Ammonia-induced cell envelope injury in Escherichia coli and Enterobacter aerogenes.
Canadian journal of microbiology (1990-08-01)
G Naundorf, N G Aumen
ABSTRACT
Ammonia-induced cell envelope injury was examined in pure cultures of Escherichia coli and Enterobacter aerogenes. Cell injury, as determined by the ratio of colony-forming units on m-T7 agar to colony-forming units on m-Endo agar, increased with exposure to increasing concentrations of ammonia. Cell envelopes appeared to be the site of injury as indicated by increasing susceptibility to lysozyme with increasing ammonia concentration. Cells exposed to ammonia also exhibited more cellular leakage than control cells. Leakage from cells exposed to ammonia included proteins, and all leaked substances increased in concentration as ammonia concentrations increased. The concentration of 2-keto-3-deoxyoctonate (KDO) in the outer membrane of E. coli increased with ammonia exposure, while KDO concentration in the outer membrane of E. aerogenes decreased. The results suggest that exposure of E. coli cells to high concentrations of ammonia disrupts the outer membrane and lipopolysaccharide-associated proteins, while E. aerogenes cells are affected through the disruption of bonds between KDO and the outer membrane.