Merck
  • Bioequivalence study of 2 formulations of film-coated tablets containing a fixed dose combination of bisoprolol fumarate 5 mg and hydrochlorothiazide 6.25 mg in healthy subjects.
Bioequivalence study of 2 formulations of film-coated tablets containing a fixed dose combination of bisoprolol fumarate 5 mg and hydrochlorothiazide 6.25 mg in healthy subjects.
Drug research (2013-03-30)
R R Tjandrawinata, E Setiawati, D A Yunaidi, I D Santoso, A Setiawati, L W Susanto
ABSTRACT
The present study was conducted to compare the bioavailability of 2 formulations of fixed-dose combination of bisoprolol fumarate 5 mg and hydrochlorotiazide (HCT) 6.25 mg film-coated tablet (test and reference formulations). This study was a randomized, single-blind, 2-period, 2-sequence cross-over study which included 18 healthy adult male and female subjects under fasting condition. The pharmacokinetic parameters, AUCt, AUCinf, Cmax, tmax, and t½ were determined based on the concentrations of bisoprolol (CAS 66722-44-9) and HCT (CAS 58-93-5), using ultra-performance liquid chromatography with tandem mass spectrometer detector (UPLC-MS/MS). In each of the 2 study periods (with a washout of 1 week) a single dose of test or reference product was administered. The geometric mean ratios (90% CI) of the test drug/reference drug for bisoprolol were 97.22% (93.75-100.83%) for AUCt(0-48), 97.20% (93.97-100.54%) for AUCinf, and 100.36% (93.83-107.34%) for Cmax; while those for HCT were 93.22% (84.72-102.57%), 93.39% (85.43-102.10%) and 99.39% (85.45-115.61%), for AUCt(0-24), AUCinf, and Cmax, respectively. The differences between the test and reference drug products for tmax values of bisoprolol as well as t½ values of both bisoprolol and hydrochlorothiazide were not statistically significant; yet, the difference was statistically significant for the tmax values of hydrochlorothiazide. There was no adverse event encountered during this bioequivalence test. It was concluded that the 2 formulations of fixed dose combination of bisoprolol fumarate 5 mg and hydrochlorotiazide (HCT) 6.25 mg film-coated tablet (the test and reference products) were bioequivalent.
Sigma-Aldrich
Bisoprolol hemifumarate salt, ≥98% (HPLC), solid
Supelco
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochlorothiazide, crystalline
Sigma-Aldrich
Hydrochlorothiazide, meets USP testing specifications
Bisoprolol for system suitability, European Pharmacopoeia (EP) Reference Standard
Bisoprolol fumarate, European Pharmacopoeia (EP) Reference Standard
Bisoprolol for peak identification, European Pharmacopoeia (EP) Reference Standard