Merck
  • Sacral radiculopathy due to cement leakage from percutaneous sacroplasty, successfully treated with surgical decompression.
Sacral radiculopathy due to cement leakage from percutaneous sacroplasty, successfully treated with surgical decompression.
Journal of neurosurgery. Spine (2013-03-26)
Sean M Barber, Andrew D Livingston, David A Cech
ABSTRACT
Percutaneous sacroplasty is a procedure adapted from vertebroplasty, which is designed to ameliorate the painful morbidity associated with sacral insufficiency fractures without the invasiveness of open surgery. Early estimates of efficacy, according to several case reports and small series, appear promising, but the procedure is not without risk. Several cases of radiculopathy due to nerve root compression by extravasated polymethylmethacrylate (PMMA) have been reported. The authors present a case of radiculopathy caused by cement leakage from sacroplasty, treated with surgical decompression of the compromised nerve root. The patient presented with left S-1 radiculopathy and was found on CT to have a left S-1 nerve root completely encased in PMMA over a portion of its length. The patient underwent sacral laminectomy with the removal of PMMA and experienced pain relief and the return of function postoperatively. Surgical removal of PMMA extravasated during sacroplasty is feasible and should be considered when nerve root compression or canal stenosis causes pain or neurological deficit refractory to conservative therapy.
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Supelco
Poly(methyl methacrylate), analytical standard, 4,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, 8,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, 20,000