Merck
  • Delayed pharyngeal repolarization promotes abnormal calcium buildup in aging muscle.
Delayed pharyngeal repolarization promotes abnormal calcium buildup in aging muscle.
Biochemical and biophysical research communications (2013-03-21)
Przemyslaw Swiatkowski, Federico Sesti
ABSTRACT
In the pharynx of Caenorhabditis elegans, the accessory subunit MPS-4, homolog to human KCNE1, forms a complex with K(+) channel EXP-2 that terminates the action potential. An aspartate residue critical for KCNE1 function, asp76, is conserved in MPS-4 (asp74). Here, we studied the effects of D74N-MPS-4 on the aging pharynx. Electrophysiological studies showed that D74N delays pharyngeal repolarization. Pharynxes of transgenic worms expressing D74N exhibited higher levels of intracellular calcium compared to normal pharynxes. Accordingly, loss of pharyngeal function was accelerated in aging D74N worms. The pharyngeal action potential resembles the action potential that controls the mechanical activity of human left ventricle. Hence, these findings argue that the hearts of patients affected by delayed repolarization, a condition known as long QT syndrome, may experience dysregulated calcium homeostasis.
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)