Merck
  • Electronic deactivation of guanosine in extended hydrogen-bonded self-assemblies.
Electronic deactivation of guanosine in extended hydrogen-bonded self-assemblies.
The journal of physical chemistry. A (2013-03-21)
Katharina Röttger, Nina K Schwalb, Friedrich Temps
ABSTRACT
Guanosine (G) derivatives in nonpolar aprotic solvents self-assemble to intricate hydrogen-bonded supramolecular architectures, including dimers, ribbons, and cyclic quartets. Considerable interest exists in the nature of the excited electronic states, their lifetimes and the radiationless deactivation mechanisms of the molecules in those environments. Here, we report on the electronic relaxation of G in the extended H-bridged networks in solution in n-hexane. The resulting architectures were sampled by FTIR, UV, and CD spectroscopies. The dynamics after 260 nm photoexcitation were investigated by femtosecond fluorescence up-conversion, broadband UV-vis absorption, and single-color deep-UV measurements. The observed temporal profiles reveal a hierarchy of relaxation processes, with lifetimes τ1 = 0.63 ± 0.03 ps, τ2 = 5.9 ± 0.3 ps, and τ3 = 62 ± 7 ps. Moreover, about 10% of the photoexcited molecules transform to much longer-lived product states with lifetime τ4 ≈ 3.6 ± 1.0 ns. These excited-state lifetimes are much longer than in the G monomer or the G·G dimers studied previously, hinting at sizable energy shifts among the excited ππ* and nπ* states and trapping of excited-state population in the supramolecular networks by potential energy barriers along the optimal electronic deactivation pathways of the molecules.
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Guanosine, BioReagent, suitable for cell culture
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane-1-13C, 99 atom % 13C
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Guanosine, ≥97.0% (HPLC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)