Merck
  • Novel riboflavin transporter family RFVT/SLC52: identification, nomenclature, functional characterization and genetic diseases of RFVT/SLC52.
Novel riboflavin transporter family RFVT/SLC52: identification, nomenclature, functional characterization and genetic diseases of RFVT/SLC52.
Molecular aspects of medicine (2013-03-20)
Atsushi Yonezawa, Ken-ichi Inui
ABSTRACT
Riboflavin, a water-soluble vitamin also known as vitamin B2, is essential for normal cellular functions. Riboflavin transporters play important roles in its homeostasis. Recently, three novel riboflavin transporters were identified, and designated as RFT1, RFT2 and RFT3. Because the RFTs did not show similarity to other SLC transporters, and RFT1 and RFT3 are similar in sequence and function, they were assigned into a new SLC family, SLC52. Subsequently, RFT1/GPR172B, RFT3/GPR172A and RFT2/C20orf54 were renamed as RFVT1/SLC52A1, RFVT2/SLC52A2 and RFVT3/SLC52A3, respectively. In this review, we summarize recent findings on the cloning, nomenclature, functional characterization and genetic diseases of RFVT1/SLC52A1, RFVT2/SLC52A2 and RFVT3/SLC52A3.
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Pricing and availability is not currently available.
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Pricing and availability is not currently available.
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Riboflavin (B2), analytical standard
Pricing and availability is not currently available.
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.