Merck
  • Preparation and characterization of pH- and temperature-responsive nanocomposite double network hydrogels.
Preparation and characterization of pH- and temperature-responsive nanocomposite double network hydrogels.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Zhiqiang Li, Jianfeng Shen, Hongwei Ma, Xin Lu, Min Shi, Na Li, Mingxin Ye
ABSTRACT
A methodology is described for the preparation of pH- and temperature-responsive double network (DN) hydrogels with poly(N-isopropylacrylamide) (PNIPAM) as a tightly crosslinked 1st network, polyacrylic acid (PAA) as a loosely crosslinked 2nd network and graphene oxide (GO) as an additive. GO sheets were first prepared via an oxidation reaction and then dispersed in NIPAM aqueous solution via silanization. Free-radical polymerization of NIPAM was carried out at 20 °C in a water bath, and then subjected to UV light, leading to the formation of pH- and temperature-responsive PNIPAM/AA/GO DN hydrogels. The effects of GO sheets and AA contents on various physical properties were investigated. Results show that PNIPAM/AA/GO hydrogels undergo a large volumetric change in response to temperature. It also exhibits significantly fast swelling/deswelling compared with conventional PNIPAM hydrogel. Moreover, the PNIPAM/AA/GO hydrogels have a much better mechanical property than the conventional PNIPAM hydrogels.
Sigma-Aldrich
Zinc acrylate, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
€124.00
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
€124.00
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
€124.00
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
€124.00
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
€124.00