Merck
  • Modeling and experimental investigations of Fano resonances in free-standing LiNbO3 photonic crystal slabs.
Modeling and experimental investigations of Fano resonances in free-standing LiNbO3 photonic crystal slabs.
Optics express (2013-03-14)
Jun Deng, Sajid Hussain, Vanga Sudheer Kumar, Wei Jia, Ching Eng Png, Lim Soon Thor, Andrew A Bettiol, Aaron J Danner
ABSTRACT
In this paper the Fano resonance in a free-standing LiNbO(3) photonic crystal slab is demonstrated. We present a numerical analysis and experimental measurements with free space illumination where the dependence of slab thickness, radius of air holes and lattice types are investigated. The unique property of polarization dependence for LiNbO(3) photonic crystal slabs is also analyzed, and we show that the transmission spectra exhibit significant sensitivity (~25nm) to polarization. A monolithic free-standing LiNbO(3) photonic crystal slab was fabricated using ion beam enhanced etching (IBEE) technology. Measurement results of the reflection spectra agree with the numerical analysis.
Product Description
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.5 mm wire