Merck
  • First report on predictive chemometric modeling, 3D-toxicophore mapping and in silico screening of in vitro basal cytotoxicity of diverse organic chemicals.
First report on predictive chemometric modeling, 3D-toxicophore mapping and in silico screening of in vitro basal cytotoxicity of diverse organic chemicals.
Toxicology in vitro : an international journal published in association with BIBRA (2013-03-14)
Supratik Kar, Kunal Roy
ABSTRACT
Classification and regression based quantitative structure-toxicity relationship (QSTR) as well as toxicophore models were developed for the first time on basal cytotoxicity data (in vitro 3T3 neutral red uptake data) of a diverse series of chemicals (including drugs and environmental pollutants) collected from the ACuteTox database (http://www.acutetox.eu/). Statistically significant QSTR models were obtained using linear discriminant analysis (classification) and partial least squares (regression) methodologies. Generated toxicophore models showed four important features responsible for basal cytotoxicity: (i) two hydrophobic aliphatic groups (HYD Aliphatic), (ii) ring aromatic group (RA) and (iii) hydrogen bond donor (HBD). The most predictive hypothesis (Hypo 1) had a correlation coefficient of 0.932 for the training set, a low rms deviation of 1.105, and an acceptable cost difference of 62.8 bits, which represents a true correlation and a good predictivity. QSTR and toxicophore models were rigorously validated internally as well as externally along with the randomization test to nullify the possibilities of chance correlation. Our in silico models enable to identify the essential structural attributes and quantify the prime molecular pre-requisites which were chiefly responsible for in vitro basal cytotoxicity. The developed models were also implemented to screen basal cytotoxicity for huge number DrugBank database (http://www.drugbank.ca/) compounds.
Sigma-Aldrich
Neutral Red, certified by the BSC
Sigma-Aldrich
Neutral Red Solution (0.33%), 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red, Dye content ≥90 %
Sigma-Aldrich
Neutral Red