Merck
Comparative glycomics of leukocyte glycosaminoglycans.
The FEBS journal (2013-03-14)
Chun Shao, Xiaofeng Shi, Mitchell White, Yu Huang, Kevan Hartshorn, Joseph Zaia
ABSTRACT
Glycosaminoglycans (GAGs) vary widely in disaccharide and oligosaccharide content in a tissue-specific manner. Nonetheless, there are common structural features, such as the presence of highly sulfated non-reducing end domains on heparan sulfate (HS) chains. Less clear are the patterns of expression of GAGs on specific cell types. Leukocytes are known to express GAGs primarily of the chondroitin sulfate (CS) type. However, little is known regarding the properties and structures of the GAG chains, their variability among normal subjects, and changes in structure associated with disease conditions. We isolated peripheral blood leukocyte populations from four human donors and extracted GAGs. We determined the relative and absolute disaccharide abundances for HS and CS GAGs classes using size exclusion chromatography-mass spectrometry (SEC-MS). We found that all leukocytes express HS chains with a level of sulfation that is more similar to heparin than to organ-derived HS. The levels of HS expression follows the trend T cells/B cells > monocytes/natural killer cells > polymorphonuclear leukocytes (PMNs). In addition, CS abundances were considerably higher than total HS but varied considerably in a leukocyte cell type-specific manner. Levels of CS were higher for myeloid lineage cells (PMNs and monocytes) than for lymphoid cells (B, T and natural killer (NK) cells). This information establishes the range of GAG structures expressed on normal leukocytes and is necessary for subsequent inquiry into disease conditions.
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Chondroitin sulfate sodium, European Pharmacopoeia (EP) Reference Standard
Chondroitin sulfate sodium (marine), European Pharmacopoeia (EP) Reference Standard