Merck
  • Remediation of nonaqueous phase liquid polluted sites using surfactant-enhanced air sparging and soil vapor extraction.
Remediation of nonaqueous phase liquid polluted sites using surfactant-enhanced air sparging and soil vapor extraction.
Water environment research : a research publication of the Water Environment Federation (2013-03-12)
Chuan-Yu Qin, Yong-Sheng Zhao, Yan Su, Wei Zheng
ABSTRACT
A two-dimensional laboratory sand tank was installed to study the remediation efficiency of surfactant-enhanced air sparging (-SEAS) coupled with soil vapor extraction (SVE) in nonaqueous phase liquid (NAPL) polluted sites. During initial stages of remediation, it was more reasonable to use conventional air sparging coupled with SVE. When most free NAPLs were removed and contaminant removal rate was maintained at a relatively low level, surfactant was added to the groundwater. During enhanced remediation, lower interfacial tension caused residual NAPLs in the porous media to slightly migrate, making the downstream contaminant concentration somewhat higher. The polluted area, however, was not more enlarged than before. The decrease in surface tension resulted in increased air saturation in the groundwater and the extent of the air influence zone. After 310 hours, 78.7% of the initial chlorobenzene mass had volatilized, 3.3% had migrated out of the sand profile, 17.5% was in the vadose zone, and 0.5% remained in the groundwater, thus revealing that SEAS/SVE can effectively improve the remediation of NAPL polluted sites.
Product Description
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%